5-Mercapto-2-methoxy-4-methylbenzoic acid ≥98%

Lieferant: Apollo Scientific

OR510234-250MG
APOSOR510234-250MGEA 540 EUR
APOSOR510234-250MG
5-Mercapto-2-methoxy-4-methylbenzoic acid ≥98%
5-Mercapto-2-methoxy-4-methylbenzoic acid
Formel: C₉H₁₀O₃S
Molecular Weight: 198,239 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09831918
CAS-Nummer: 439579-12-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR