6-Bromo-[1,2,4]triazolo[4,3-a]pyrimidine ≥98%

Lieferant: Apollo Scientific

OR510235-1G
APOSOR510235-1GEA 230 EUR
APOSOR510235-1G
6-Bromo-[1,2,4]triazolo[4,3-a]pyrimidine ≥98%
6-Bromo-[1,2,4]triazolo[4,3-a]pyrimidine
Formel: C₅H₃BrN₄
Molecular Weight: 199,008 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09994391
CAS-Nummer: 1019023-12-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR