(5-Chlorobenzo[d][1,3]dioxol-4-yl)boronic acid ≥98%

Lieferant: Apollo Scientific

Synonyme: (5-Chlorobenzo[d][1,3]dioxol-4-yl)boronicacid

OR510236-100MG
APOSOR510236-100MGEA 46.4 EUR
APOSOR510236-100MG
(5-Chlorobenzo[d][1,3]dioxol-4-yl)boronic acid ≥98%
(5-Chlorobenzo[d][1,3]dioxol-4-yl)boronic acid
Formel: C₇H₆BClO₄
Molecular Weight: 200,384 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31382028
CAS-Nummer: 2087452-50-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR