5-Bromo-3-methylquinoline ≥98%
Lieferant: Apollo Scientific
OR510238-1G
APOSOR510238-1GEA
505
EUR
APOSOR510238-1G
5-Bromo-3-methylquinoline ≥98%
5-Bromo-3-methylquinoline
Formel:
C₁₀H₈BrN Molecular Weight: 222,081 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23701524 CAS-Nummer: 397322-46-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...