2-Bromo-5-iodoimidazo[2,1-b]thiazole ≥98%
Lieferant: Apollo Scientific
OR510240-100MG
APOSOR510240-100MGEA
195
EUR
APOSOR510240-100MG
2-Bromo-5-iodoimidazo[2,1-b]thiazole ≥98%
2-Bromo-5-iodoimidazo[2,1-b]thiazole
Formel:
C₅H₂BrIN₂S Molecular Weight: 328,956 g/mol Lagertemperatur: Raumtemperatur |
CAS-Nummer:
1379307-48-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...