4-Amino-2-methylpyrimidine-5-carboxylic acid ≥98%

Lieferant: Apollo Scientific

OR510241-250MG
APOSOR510241-250MGEA 40.6 EUR
APOSOR510241-250MG
4-Amino-2-methylpyrimidine-5-carboxylic acid ≥98%
4-Amino-2-methylpyrimidine-5-carboxylic acid
Formel: C₆H₇N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00186151
CAS-Nummer: 769-52-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR