Quinoline-3,4-diamine ≥98%

Lieferant: Apollo Scientific

OR510242-100MG
APOSOR510242-100MGEA 440 EUR
APOSOR510242-100MG
Quinoline-3,4-diamine ≥98%
Quinoline-3,4-diamine
Formel: C₉H₉N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00666954
CAS-Nummer: 87751-33-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR