3-Formyl-2-hydroxybenzoic acid ≥98%

Lieferant: Apollo Scientific

OR510243-250MG
APOSOR510243-250MGEA 389 EUR
APOSOR510243-250MG
3-Formyl-2-hydroxybenzoic acid ≥98%
3-Formyl-2-hydroxybenzoic acid
Formel: C₈H₆O₄
Molecular Weight: 166,131 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00143165
CAS-Nummer: 610-04-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR