1,7-Naphthyridine-3-carboxylic acid ≥98%

Lieferant: Apollo Scientific

OR510245-100MG
APOSOR510245-100MGEA 343 EUR
APOSOR510245-100MG
1,7-Naphthyridine-3-carboxylic acid ≥98%
1,7-Naphthyridine-3-carboxylic acid
Formel: C₉H₆N₂O₂
Molecular Weight: 174,156 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11044184
CAS-Nummer: 250674-49-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR