Methyl 3-amino-4-methyl-2-nitrobenzoate ≥98%

Lieferant: Apollo Scientific

OR510247-100MG
APOSOR510247-100MGEA 249 EUR
APOSOR510247-100MG
Methyl 3-amino-4-methyl-2-nitrobenzoate ≥98%
Methyl 3-amino-4-methyl-2-nitrobenzoate
Formel: C₉H₁₀N₂O₄
Molecular Weight: 210,187 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28131934
CAS-Nummer: 1824056-12-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR