Methyl 4-bromo-6-cyanobenzo[b]thiophene-2-carboxylate ≥98%
Lieferant: Apollo Scientific
OR510249-100MG
APOSOR510249-100MGEA
347
EUR
APOSOR510249-100MG
Methyl 4-bromo-6-cyanobenzo[b]thiophene-2-carboxylate ≥98%
Methyl 4-bromo-6-cyanobenzo[b]thiophene-2-carboxylate
Formel:
C₁₁H₆BrNO₂S Molecular Weight: 296,14 g/mol Lagertemperatur: Raumtemperatur |
CAS-Nummer:
753455-45-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...