tert-Butyl 3-bromo-1,4,6,7-tetrahydro-5H-pyrazolo[4,3-c]pyridine-5-carboxylate ≥98%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 3-bromo-6,7-dihydro-1H-pyrazolo[4,3-c]pyridine-5(4H)-carboxylate

OR510250-100MG
APOSOR510250-100MGEA 128 EUR
APOSOR510250-100MG
tert-Butyl 3-bromo-1,4,6,7-tetrahydro-5H-pyrazolo[4,3-c]pyridine-5-carboxylate ≥98%
tert-Butyl 3-bromo-1,4,6,7-tetrahydro-5H-pyrazolo[4,3-c]pyridine-5-carboxylate
Formel: C₁₁H₁₆BrN₃O₂
Molecular Weight: 302,168 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30751004
CAS-Nummer: 1936429-06-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR