(R)-AntPhos ≥98%

Lieferant: Apollo Scientific

Synonyme: (R)-4-(Anthracen-9-yl)-3-(tert-butyl)-2,3-dihydrobenzo[d][1,3]oxaphosphole

OR510251-100MG
APOSOR510251-100MGEA 98.6 EUR
APOSOR510251-100MG
(R)-AntPhos ≥98%
(R)-AntPhos
Formel: C₂₅H₂₃OP
Molecular Weight: 370,423 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630706
CAS-Nummer: 1456816-37-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR