(S)-AntPhos ≥98%

Lieferant: Apollo Scientific

Synonyme: (S)-4-(Anthracen-9-yl)-3-(tert-butyl)-2,3-dihydrobenzo[d][1,3]oxaphosphole

OR510252-100MG
APOSOR510252-100MGEA 85.7 EUR
APOSOR510252-100MG
(S)-AntPhos ≥98%
(S)-AntPhos
Formel: C₂₅H₂₃OP
Molecular Weight: 370,423 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630706
CAS-Nummer: 1807740-34-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR