(S)-BI-DIME ≥98%

Lieferant: Apollo Scientific

Synonyme: (S)-3-(tert-Butyl)-4-(2,6-dimethoxyphenyl)-2,3-dihydrobenzo[d][1,3]oxaphosphole

OR510254-100MG
APOSOR510254-100MGEA 56.7 EUR
APOSOR510254-100MG
(S)-BI-DIME ≥98%
(S)-BI-DIME
Formel: C₁₉H₂₃O₃P
Molecular Weight: 330,358 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630704
CAS-Nummer: 1373432-09-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR