(S,S)-Me-BI-DIME ≥98%
Lieferant: Apollo Scientific
Synonyme:
(2S,3S)-3-(tert-Butyl)-4-(2,6-dimethoxyphenyl)-2-methyl-2,3-dihydrobenzo[d][1,3]oxaphosphole
OR510255-100MG
APOSOR510255-100MGEA
296
EUR
APOSOR510255-100MG
(S,S)-Me-BI-DIME ≥98%
(S,S)-Me-BI-DIME
Formel:
C₂₀H₂₅O₃P Molecular Weight: 344,384 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31707596 CAS-Nummer: 1373432-11-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...