(S,S)-iPr-BI-DIME ≥98%

Lieferant: Apollo Scientific

Synonyme: (2S,3S)-3-(tert-Butyl)-4-(2,6-dimethoxyphenyl)-2-isopropyl-2,3-dihydrobenzo[d][1,3]oxaphosphole

OR510257-100MG
APOSOR510257-100MGEA 116 EUR
APOSOR510257-100MG
(S,S)-iPr-BI-DIME ≥98%
(S,S)-iPr-BI-DIME
Formel: C₂₂H₂₉O₃P
Molecular Weight: 372,438 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32206571
CAS-Nummer: 1477517-21-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR