(2R,2'R,3R,3'R)-WingPhos ≥98%

Lieferant: Apollo Scientific

Synonyme: (2R,2'R,3R,3'R)-4,4'-Di(anthracen-9-yl)-3,3'-di-tert-butyl-2,2',3,3'-tetrahydro-2,2'-bibenzo[d][1,3]oxaphosphole

OR510258-100MG
APOSOR510258-100MGEA 132 EUR
APOSOR510258-100MG
(2R,2'R,3R,3'R)-WingPhos ≥98%
(2R,2'R,3R,3'R)-WingPhos
Formel: C₅₀H₄₄O₂P₂
Molecular Weight: 738,831 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31630707
CAS-Nummer: 1884680-45-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR