(2S,2'S,3S,3'S)-WingPhos ≥98%
Lieferant: Apollo Scientific
Synonyme:
(2S,2'S,3S,3'S)-4,4'-Di(anthracen-9-yl)-3,3'-di-tert-butyl-2,2',3,3'-tetrahydro-2,2'-bibenzo[d][1,3]oxaphosphole
OR510259-100MG
APOSOR510259-100MGEA
122
EUR
APOSOR510259-100MG
(2S,2'S,3S,3'S)-WingPhos ≥98%
(2S,2'S,3S,3'S)-WingPhos
Formel:
C₅₀H₄₄O₂P₂ Molecular Weight: 738,831 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31630708 CAS-Nummer: 1435940-19-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...