8H-Indolo[3,2,1-de]acridin-8-one 99%

Lieferant: Apollo Scientific

OR51026-1G
APOSOR51026-1GEA 446 EUR
APOSOR51026-1G
8H-Indolo[3,2,1-de]acridin-8-one 99%
8H-Indolo[3,2,1-de]acridin-8-one
Formel: C₁₉H₁₁NO
Molecular Weight: 269.297 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 32081-26-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR