2-Bromo-5-methoxypyridin-4-amine ≥98%

Lieferant: Apollo Scientific

OR510260-100MG
APOSOR510260-100MGEA 56.7 EUR
APOSOR510260-100MG
2-Bromo-5-methoxypyridin-4-amine ≥98%
2-Bromo-5-methoxypyridin-4-amine
Formel: C₆H₇BrN₂O
Molecular Weight: 203,037 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26520651
CAS-Nummer: 1417644-40-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR