6-Bromoimidazo[1,5-a]pyridine-1-carboxylic acid ≥98%

Lieferant: Apollo Scientific

OR510261-100MG
APOSOR510261-100MGEA 654 EUR
APOSOR510261-100MG
6-Bromoimidazo[1,5-a]pyridine-1-carboxylic acid ≥98%
6-Bromoimidazo[1,5-a]pyridine-1-carboxylic acid
Formel: C₈H₅BrN₂O₂
Molecular Weight: 241,042 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1427405-61-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR