3-Iodo-6-methyl-1H-pyrazolo[3,4-d]pyrimidine ≥98%

Lieferant: Apollo Scientific

OR510262-100MG
APOSOR510262-100MGEA 565 EUR
APOSOR510262-100MG
3-Iodo-6-methyl-1H-pyrazolo[3,4-d]pyrimidine ≥98%
3-Iodo-6-methyl-1H-pyrazolo[3,4-d]pyrimidine
Formel: C₆H₅IN₄
Molecular Weight: 260,035 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1415559-47-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR