5,7-Dibromobenzo[d]thiazole ≥98%

Lieferant: Apollo Scientific

OR510263-100MG
APOSOR510263-100MGEA 212 EUR
APOSOR510263-100MG
5,7-Dibromobenzo[d]thiazole ≥98%
5,7-Dibromobenzo[d]thiazole
Formel: C₇H₃Br₂NS
Molecular Weight: 292,978 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12962849
CAS-Nummer: 875-69-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR