5-Amino-2-chloropyrimidine-4-carboxylic acid ≥98%

Lieferant: Apollo Scientific

OR510264-100MG
APOSOR510264-100MGEA 142 EUR
APOSOR510264-100MG
5-Amino-2-chloropyrimidine-4-carboxylic acid ≥98%
5-Amino-2-chloropyrimidine-4-carboxylic acid
Formel: C₅H₄ClN₃O₂
Molecular Weight: 173,557 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20040107
CAS-Nummer: 1052714-41-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR