(2S)-1,2-Propandiaminhydrochlorid

Lieferant: Apollo Scientific

OR51195-100G
APOSOR51195-100GEA 150 EUR
APOSOR51195-100G
(2S)-1,2-Propandiaminhydrochlorid
(S)-Propane-1,2-diamine dihydrochloride
Formel: C₃H₁₂Cl₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00050706
CAS-Nummer: 19777-66-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR