tert-Butyl N-amino-N-[(2-bromophenyl)methyl]carbamate 95%

Lieferant: Apollo Scientific

OR51363-1G OR51363-250MG
APOSOR51363-1GEA 609 EUR
APOSOR51363-1G APOSOR51363-250MG
tert-Butyl N-amino-N-[(2-bromophenyl)methyl]carbamate 95%
tert-Butyl N-amino-N-[(2-bromophenyl)methyl]carbamate
Formel: C₁₂H₁₇BrN₂O₂
Molecular Weight: 301.18 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 2268038-58-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR