3-Bromo-4-hydroxy-5-methoxyphenylacetic acid

Lieferant: Apollo Scientific
Achtung

OR5143-250MG
APOSOR5143-250MGEA 0 EUR
APOSOR5143-250MG
3-Bromo-4-hydroxy-5-methoxyphenylacetic acid
3-Bromo-4-hydroxy-5-methoxyphenylacetic acid
Formel: C₉H₉BrO₄
Schmelzpunkt: 193…195 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00060354
CAS-Nummer: 206559-42-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR