1-Benzyl-3-phenylpiperazine

Lieferant: Apollo Scientific

OR5145-1G
APOSOR5145-1GEA 321 EUR
APOSOR5145-1G
1-Benzyl-3-phenylpiperazine
1-Benzyl-3-phenylpiperazine
Formel: C₁₇H₂₀N₂
Molecular Weight: 252,354 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07367740
CAS-Nummer: 5368-32-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR