2-Acetamido-2-deoxy-4-O-(α-L-fucopyranosyl)-D-glucopyranose
Lieferant: Apollo Scientific
Synonyme:
Fuc1-a-4GlcNAc
OR5155T-5MG
APOSOR5155T-5MGEA
321
EUR
APOSOR5155T-5MG
2-Acetamido-2-deoxy-4-O-(α-L-fucopyranosyl)-D-glucopyranose
2-Acetamido-2-deoxy-4-O-(α-L-fucopyranosyl)-D-glucopyranose
Formel:
C₁₄H₂₅NO₁₀ Schmelzpunkt: 216…218 °C (Zers.) Lagertemperatur: Gefrierschrank |
MDL:
MFCD00153897 CAS-Nummer: 76211-71-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...