3-Amino-N-methylpropanamide hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
OR51734-500MG
APOSOR51734-500MGEA
224
EUR
APOSOR51734-500MG
3-Amino-N-methylpropanamide hydrochloride
3-Amino-N-methylpropanamide hydrochloride
Formel:
C₄H₁₁ClN₂O Molecular Weight: 138.6 g/mol Schmelzpunkt: 110…112 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD10686841 CAS-Nummer: 51739-61-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...