2-Amino-5-(3-nitrophenyl)-1,3,4-thiadiazole 95+%

Lieferant: Apollo Scientific

Synonyme: 5-(3-Nitrophenyl)-1,3,4-thiadiazol-2-amine, 3-(5-Amino-1,3,4-thiadiazol-2-yl)nitrobenzene

OR51736-1G
APOSOR51736-1GEA 68.3 EUR
APOSOR51736-1G
2-Amino-5-(3-nitrophenyl)-1,3,4-thiadiazole 95+%
2-Amino-5-(3-nitrophenyl)-1,3,4-thiadiazole
Formel: C₈H₆N₄O₂S
Molecular Weight: 222,224 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00453655
CAS-Nummer: 833-47-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR