1-(2-Methoxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-[1-(2-Methoxyethyl)-1H-pyrazol-4-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-(2-Methoxyethyl)-1H-pyrazole-4-boronic acid Pinacol ester, 1-(2-Methoxyethyl)-1H-pyrazole-4-boronic acid, pinacol ester
OR51737-250MG
APOSOR51737-250MGEA
30.6
EUR
APOSOR51737-250MG
1-(2-Methoxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol
1-(2-Methoxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Formel:
C₁₂H₂₁BN₂O₃ Molecular Weight: 252,1177 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD12405649 CAS-Nummer: 847818-71-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...