1-(2-Hydroxyethyl)-1H-pyrazole-4-boronic acid, pinacol ester 95%

Lieferant: Apollo Scientific
Achtung

OR51738-250MG OR51738-5G
APOSOR51738-250MGEA 63.8 EUR
APOSOR51738-250MG APOSOR51738-5G
1-(2-Hydroxyethyl)-1H-pyrazole-4-boronic acid, pinacol ester 95%
1-(2-Hydroxyethyl)-1H-pyrazole-4-boronic acid, pinacol ester
Formel: C₁₁H₁₉BN₂O₃
Lagertemperatur: Gefrierschrank
MDL: MFCD12033564
CAS-Nummer: 1040377-08-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR