1-Propylpiperazine hydrochloride

Lieferant: Apollo Scientific

Synonyme: 1-(Piperazin-1-yl)propane hydrochloride

OR51746-50MG
APOSOR51746-50MGEA 31.9 EUR
APOSOR51746-50MG
1-Propylpiperazine hydrochloride
1-Propylpiperazine hydrochloride
Formel: C₇H₁₆N₂·ClH
Molecular Weight: 164,676 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11575790
CAS-Nummer: 71888-55-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR