(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl (2S,4R)-4-hydroxy-2-hydroxymethylpyrrolidine-1-carboxylate, N-Boc-trans-4-hydroxy-L-prolinol, (2S,4R)-1-Boc-4-hydroxy-2-(hydroxymethyl)pyrrolidine, Boc-Hyp-OL

OR51749-25G
APOSOR51749-25GEA 97.3 EUR
APOSOR51749-25G
(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine 96%
(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxy-2-(hydroxymethyl)pyrrolidine
Formel: C₁₀H₁₉NO₄
Molecular Weight: 217,27 g/mol
Schmelzpunkt: 81…86 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02094386
CAS-Nummer: 61478-26-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR