2-Acetyl-5-brompyridin

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(5-Bromopyridin-2-yl)ethanone

OR51752-10G OR51752-1G OR51752-5G
APOSOR51752-10GEA 81.2 EUR
APOSOR51752-10G APOSOR51752-1G APOSOR51752-5G
2-Acetyl-5-brompyridin
2-Acetyl-5-brompyridin
Formel: C₇H₆BrNO
Molecular Weight: 200.0300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04974523
CAS-Nummer: 214701-49-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR