2-Acetyl-5-brompyridin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(5-Bromopyridin-2-yl)ethanone
OR51752-10G
OR51752-1G
OR51752-5G
APOSOR51752-10GEA
81.2
EUR
APOSOR51752-10G
APOSOR51752-1G
APOSOR51752-5G
2-Acetyl-5-brompyridin
2-Acetyl-5-brompyridin
Formel:
C₇H₆BrNO Molecular Weight: 200.0300 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04974523 CAS-Nummer: 214701-49-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...