3-chloro-4-hydroxypyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chloropyridin-4-ol

OR51765-250MG
APOSOR51765-250MGEA 21.9 EUR
APOSOR51765-250MG
3-chloro-4-hydroxypyridine
3-chloro-4-hydroxypyridine
Formel: C₅H₄ClNO
Molecular Weight: 129,543 g/mol
Schmelzpunkt: 204 - 205 °C
MDL: MFCD04114248
CAS-Nummer: 89284-20-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR