4,6-Dichloropyridin-3-ol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,4-Dichloro-5-hydroxypyridine

OR51775-1G
APOSOR51775-1GEA 393 EUR
APOSOR51775-1G
4,6-Dichloropyridin-3-ol
4,6-Dichloropyridin-3-ol
Formel: C₅H₃Cl₂NO
Molecular Weight: 163,99 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13190072
CAS-Nummer: 1196157-47-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR