4-Hydroxy-3-methoxypyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Methoxypyridin-4-ol

OR51777-1G
APOSOR51777-1GEA 293 EUR
APOSOR51777-1G
4-Hydroxy-3-methoxypyridine
4-Hydroxy-3-methoxypyridine
Formel: C₆H₇NO₂
Molecular Weight: 125,125 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16998801
CAS-Nummer: 62885-41-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR