2-Bromo-4-hydroxy-5-methoxypyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Bromo-4-hydroxypyridin-3-yl methyl ether, 2-Bromo-5-methoxypyridin-4-ol

OR51781-1G
APOSOR51781-1GEA 194 EUR
APOSOR51781-1G
2-Bromo-4-hydroxy-5-methoxypyridine
2-Bromo-4-hydroxy-5-methoxypyridine
Formel: C₆H₆BrNO₂
Molecular Weight: 204,021 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20327975
CAS-Nummer: 1344046-11-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR