4-Methoxypyridin-3-ol
Lieferant: Apollo Scientific
|
Achtung
|
OR51782-1G
OR51782-250MG
OR51782-100MG
APOSOR51782-1GEA
186
EUR
APOSOR51782-1G
APOSOR51782-250MG
APOSOR51782-100MG
4-Methoxypyridin-3-ol
4-Methoxypyridin-3-ol
Formel:
C₆H₇NO₂ Schmelzpunkt: 143…144 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11044354 CAS-Nummer: 153199-54-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...