2-Chloro-5-hydroxy-4-methylpyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Chloro-4-methylpyridin-3-ol, 2-Chloro-5-hydroxy-4-picoline

OR51783-1G
APOSOR51783-1GEA 379 EUR
APOSOR51783-1G
2-Chloro-5-hydroxy-4-methylpyridine
2-Chloro-5-hydroxy-4-methylpyridine
Formel: C₆H₆ClNO
Molecular Weight: 143,571 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16610471
CAS-Nummer: 1227502-89-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR