5-Bromo-3-hydroxy-2-methoxypyridine

Lieferant: Apollo Scientific
Achtung

OR51784-100MG OR51784-500MG
APOSOR51784-100MGEA 87 EUR
APOSOR51784-100MG APOSOR51784-500MG
5-Bromo-3-hydroxy-2-methoxypyridine
5-Bromo-3-hydroxy-2-methoxypyridine
Formel: C₆H₆BrNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18256269
CAS-Nummer: 1211589-04-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR