(R)-(-)-3-Isopropyl-2,5-piperazindion
Lieferant: Apollo Scientific
|
Achtung
|
OR51793-5G
APOSOR51793-5GEA
120
EUR
APOSOR51793-5G
(R)-(-)-3-Isopropyl-2,5-piperazindion
(R)-(-)-3-Isopropyl-2,5-piperazindion
Formel:
C₇H₁₂N₂O₂ Schmelzpunkt: 256…262 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00210060 CAS-Nummer: 143673-66-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...