(3,5-Dichlorphenyl)essigsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(3,5-Dichlorophenyl)acetic acid

OR51794-5G
APOSOR51794-5GEA 26.1 EUR
APOSOR51794-5G
(3,5-Dichlorphenyl)essigsäure
(3,5-Dichlorphenyl)essigsäure
Formel: C₈H₆Cl₂O₂
Molecular Weight: 205,03 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD01764671
CAS-Nummer: 51719-65-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR