(S)-2-Piperidinone-6-carboxylic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2S)-6-Oxopiperidine-2-carboxylic acid

OR51818-1G
APOSOR51818-1GEA 58 EUR
APOSOR51818-1G
(S)-2-Piperidinone-6-carboxylic acid 98%
(S)-2-Piperidinone-6-carboxylic acid
Formel: C₆H₉NO₃
Molecular Weight: 143.1406 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03839863
CAS-Nummer: 34622-39-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR