5-Chloroquinaldine 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Chloro-2-methylquinoline
OR51848-1G
APOSOR51848-1GEA
243
EUR
APOSOR51848-1G
5-Chloroquinaldine 98%
5-Chloroquinaldine
Formel:
C₁₀H₈ClN Molecular Weight: 177.63 g/mol Siedepunkt: 276…278 °C Schmelzpunkt: 25…30 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07802817 CAS-Nummer: 4964-69-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...