1-(Bromomethyl)cyclopropane-1-carbonitrile

Lieferant: Apollo Scientific
Achtung

OR51852-1G
APOSOR51852-1GEA 0 EUR
APOSOR51852-1G
1-(Bromomethyl)cyclopropane-1-carbonitrile
1-(Bromomethyl)cyclopropane-1-carbonitrile
Formel: C₅H₆BrN
Lagertemperatur: Kühlschrank
MDL: MFCD16875700
CAS-Nummer: 98730-78-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR